3-(Bromomethyl)pyridazine hydrobromide
Supplier: Apollo Scientific
OR50997-1G OR50997-250MG
APOSOR50997-1GEA 474 EUR
APOSOR50997-1G APOSOR50997-250MG
3-(Bromomethyl)pyridazine hydrobromide
3-(Bromomethyl)pyridazine hydrobromide
Formula: C₅H₅BrN₂·BrH
MW: 253.923 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20526346
CAS Number: 1452483-94-1