2-Chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane 2-oxide 98%
Supplier: Apollo Scientific
OR50999-25G OR50999-5G OR50999-100G
APOSOR50999-25GEA 89.9 EUR
APOSOR50999-25G APOSOR50999-5G APOSOR50999-100G
2-Chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane 2-oxide 98%
2-Chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane 2-oxide
Formula: C₅H₁₀ClO₃P
MW: 184.558 g/mol
Melting Pt: 97…102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03425416
CAS Number: 4090-55-5
UN: 1759
ADR: 8,III