5-Nitropyridin-3-amine hydrochloride ≥98%
Supplier: Apollo Scientific
OR510092-1G OR510092-250MG
APOSOR510092-1GEA 116 EUR
APOSOR510092-1G APOSOR510092-250MG
5-Nitropyridin-3-amine hydrochloride ≥98%
5-Nitropyridin-3-amine hydrochloride
Formula: C₅H₅N₃O₂·ClH
MW: 175.573 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15474982
CAS Number: 1220040-21-0