8H-Indolo[3,2,1-de]acridin-8-one 99%

Supplier: Apollo Scientific

OR51026-1G
APOSOR51026-1GEA 446 EUR
APOSOR51026-1G
8H-Indolo[3,2,1-de]acridin-8-one 99%
8H-Indolo[3,2,1-de]acridin-8-one
Formula: C₁₉H₁₁NO
MW: 269.297 g/mol
Storage Temperature: Ambient
CAS Number: 32081-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR