Disodium 7-aminonaphthalene-1,3-disulphonate 80%
Supplier: Apollo Scientific
Warning
Synonyms: 7-Aminonaphthalene-1,3-disulphonic acid disodium salt
OR5155-100G OR5155-250G OR5155-500G
APOSOR5155-100GEA 92.8 EUR
APOSOR5155-100G APOSOR5155-250G APOSOR5155-500G
Disodium 7-aminonaphthalene-1,3-disulphonate 80%
Disodium 7-aminonaphthalene-1,3-disulphonate
Formula: C₁₀H₇NO₆S₂·2Na
Storage Temperature: Ambient
MDL Number: MFCD07773075
CAS Number: 842-17-1