3-(Phenylethynyl)benzaldehyde 95%
Supplier: Apollo Scientific
OR51556-1G OR51556-250MG
APOSOR51556-1GEA 864 EUR
APOSOR51556-1G APOSOR51556-250MG
3-(Phenylethynyl)benzaldehyde 95%
3-(Phenylethynyl)benzaldehyde
Formula: C₁₅H₁₀O
MW: 206.239 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02736017
CAS Number: 115021-39-1