2-Ethoxy-3-hydroxypyridine
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 3-hydroxypyridin-2-yl ether, 2-Ethoxypyridin-3-ol
OR51768-1G OR51768-500MG
APOSOR51768-1GEA 367 EUR
APOSOR51768-1G APOSOR51768-500MG
2-Ethoxy-3-hydroxypyridine
2-Ethoxy-3-hydroxypyridine
Formula: C₇H₉NO₂
MW: 139.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09750035
CAS Number: 188670-05-5