4,6-Dimethoxy-1,3,5-triazin-2(1H)-one 95%
Supplier: Apollo Scientific
Warning
OR51819-1G OR51819-5G OR51819-10G
APOSOR51819-1GEA 87 EUR
APOSOR51819-1G APOSOR51819-5G APOSOR51819-10G
4,6-Dimethoxy-1,3,5-triazin-2(1H)-one 95%
4,6-Dimethoxy-1,3,5-triazin-2(1H)-one
Formula: C₅H₇N₃O₃
MW: 157.127 g/mol
Boiling Pt: 360.6 °C
Flash Pt: 171.9 °C
Storage Temperature: Freezer
MDL Number: MFCD02932731
CAS Number: 1075-59-8