2,4,5-Trichloropyridine 98%
Supplier: Apollo Scientific
Warning
OR51890-1G OR51890-5G
APOSOR51890-1GEA 82.9 EUR
APOSOR51890-1G APOSOR51890-5G
2,4,5-Trichloropyridine 98%
2,4,5-Trichloropyridine
Formula: C₅H₂Cl₃N
MW: 182.435 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09028037
CAS Number: 55934-01-5