2-Amino-5-bromo-1,3-thiazole 97%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromothiazol-2-amine
OR52147-25G OR52147-5G
APOSOR52147-25GEA 169 EUR
APOSOR52147-25G APOSOR52147-5G
2-Amino-5-bromo-1,3-thiazole 97%
2-Amino-5-bromo-1,3-thiazole
Formula: C₃H₃BrN₂S
MW: 179.038 g/mol
Melting Pt: 165 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00016893
CAS Number: 3034-22-8