(1H-Pyrazol-3-yl)methanol
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Hydroxymethyl)pyrazole
OR52395-1G OR52395-5G
APOSOR52395-1GEA 65.7 EUR
APOSOR52395-1G APOSOR52395-5G
(1H-Pyrazol-3-yl)methanol
(1H-Pyrazol-3-yl)methanol
Formula: C₄H₆N₂O
MW: 98.103 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08436148
CAS Number: 23585-49-1