5,6,7,8-Tetrahydro-2-quinolinylmethanamine 95%
Supplier: Apollo Scientific
Warning
OR52425-1G OR52425-250MG
APOSOR52425-1GEA 1160 EUR
APOSOR52425-1G APOSOR52425-250MG
5,6,7,8-Tetrahydro-2-quinolinylmethanamine 95%
5,6,7,8-Tetrahydro-2-quinolinylmethanamine
Formula: C₁₀H₁₄N₂
MW: 162.232 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12401090
CAS Number: 1351972-02-5