2-Chlorothieno[3,2-d]pyrimidin-4(3H)-one 97%
Supplier: Apollo Scientific
OR52432-1G OR52432-250MG
APOSOR52432-1GEA 68.3 EUR
APOSOR52432-1G APOSOR52432-250MG
2-Chlorothieno[3,2-d]pyrimidin-4(3H)-one 97%
2-Chlorothieno[3,2-d]pyrimidin-4(3H)-one
Formula: C₆H₃ClN₂OS
MW: 186.619 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19382468
CAS Number: 1245811-20-4