1-Methyl-1H-imidazol-2-amine 97%
Supplier: Apollo Scientific
Warning
OR52567-1G OR52567-250MG
APOSOR52567-1GEA 364 EUR
APOSOR52567-1G APOSOR52567-250MG
1-Methyl-1H-imidazol-2-amine 97%
1-Methyl-1H-imidazol-2-amine
Formula: C₄H₇N₃
MW: 97.118 g/mol
Melting Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01646103
CAS Number: 6646-51-1