1-Methyl-1H-pyrazole-4,5-diamine sulphate 97%
Supplier: Apollo Scientific
Warning
OR52687-1G OR52687-250MG
APOSOR52687-1GEA 157 EUR
APOSOR52687-1G APOSOR52687-250MG
1-Methyl-1H-pyrazole-4,5-diamine sulphate 97%
1-Methyl-1H-pyrazole-4,5-diamine sulphate
Formula: C₄H₈N₄·H₂SO₄
MW: 210.212 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28099330
CAS Number: 20055-01-0