2-Amino-4-hydroxybenzoic acid 97%
Supplier: Apollo Scientific
OR52786-1G OR52786-250MG
APOSOR52786-1GEA 267 EUR
APOSOR52786-1G APOSOR52786-250MG
2-Amino-4-hydroxybenzoic acid 97%
2-Amino-4-hydroxybenzoic acid
Formula: C₇H₇NO₃
MW: 153.135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00071506
CAS Number: 38160-63-3