Methyl 2,3,5-triiodobenzoate
Supplier: Apollo Scientific
OR52916-1G
OR52916-5G
APOSOR52916-1GEA
111
EUR
APOSOR52916-1G
APOSOR52916-5G
Methyl 2,3,5-triiodobenzoate
Methyl 2,3,5-triiodobenzoate
Formula:
C₈H₅I₃O₂ Boiling Pt: 204…244 °C Melting Pt: 104…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00463862 CAS Number: 15396-41-5 |