6-Methylpyridazin-4-amine hydrochloride
Supplier: Apollo Scientific
Warning
OR52976-1G OR52976-250MG OR52976-5G
APOSOR52976-1GEA 522 EUR
APOSOR52976-1G APOSOR52976-250MG APOSOR52976-5G
6-Methylpyridazin-4-amine hydrochloride
6-Methylpyridazin-4-amine hydrochloride
Formula: C₅H₇N₃·ClH
MW: 145.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29907144
CAS Number: 1892936-61-6