3-Chloro-4-ethynylpyridine 97%
Supplier: Apollo Scientific
Warning
Synonyms: (3-Chloropyridin-4-yl)acetylene
OR53012-250MG OR53012-1G
APOSOR53012-250MGEA 201 EUR
APOSOR53012-250MG APOSOR53012-1G
3-Chloro-4-ethynylpyridine 97%
3-Chloro-4-ethynylpyridine
Formula: C₇H₄ClN
MW: 137.566 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189773
CAS Number: 1196156-71-4