6-Ethynylimidazo[1,2-a]pyrazine
Supplier: Apollo Scientific
Warning
Synonyms: (Imidazo[1,2-a]pyrazin-6-yl)acetylene
OR53013-1G OR53013-250MG
APOSOR53013-1GEA 0 EUR
APOSOR53013-1G APOSOR53013-250MG
6-Ethynylimidazo[1,2-a]pyrazine
6-Ethynylimidazo[1,2-a]pyrazine
Formula: C₈H₅N₃
MW: 143.145 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21647858
CAS Number: 1372096-41-7