6-Bromo-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole hydrochloride 98%
Supplier: Apollo Scientific
Warning
OR53024-1G OR53024-5G OR53024-10G
APOSOR53024-1GEA 273 EUR
APOSOR53024-1G APOSOR53024-5G APOSOR53024-10G
6-Bromo-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole hydrochloride 98%
6-Bromo-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole hydrochloride
Formula: C₁₁H₁₁BrN₂·ClH
MW: 287.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12913162
CAS Number: 1059630-11-3