2-Methyl-1,4-phenylene bis{4-[3-(acryloyloxy)propoxy]benzoate}
Supplier: Apollo Scientific
Warning
Synonyms: 1,4-Bis-[4-(3-acryloyloxypropyloxy)benzoyloxy]-2-methylbenzene
OR53135-1G OR53135-5G OR53135-25G
APOSOR53135-1GEA 43.5 EUR
APOSOR53135-1G APOSOR53135-5G APOSOR53135-25G
2-Methyl-1,4-phenylene bis{4-[3-(acryloyloxy)propoxy]benzoate}
2-Methyl-1,4-phenylene bis{4-[3-(acryloyloxy)propoxy]benzoate}
Formula: C₃₃H₃₂O₁₀
MW: 588.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09743681
CAS Number: 174063-87-7