cis-Hexahydroisoindole hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: cis-Octahydro-1H-isoindole hydrochloride
OR53183-1G OR53183-5G
APOSOR53183-1GEA 50.9 EUR
APOSOR53183-1G APOSOR53183-5G
cis-Hexahydroisoindole hydrochloride
cis-Hexahydroisoindole hydrochloride
Formula: C₈H₁₅N·ClH
MW: 161.672 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09026400
CAS Number: 161829-92-1