2-(2,5-Dichlorophenyl)-1,4-benzoquinone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2',5'-Dichloro-[1,1'-biphenyl]-2,5-dione, TPI-1
OR53217-50MG OR53217-100MG OR53217-10MG
APOSOR53217-50MGEA 50.9 EUR
APOSOR53217-50MG APOSOR53217-100MG APOSOR53217-10MG
2-(2,5-Dichlorophenyl)-1,4-benzoquinone 98%
2-(2,5-Dichlorophenyl)-1,4-benzoquinone
Formula: C₁₂H₆Cl₂O₂
MW: 253.081 g/mol
Melting Pt: 114…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00448731
CAS Number: 79756-69-7