3'-Bromo-2'-hydroxyacetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3-Bromo-2-hydroxyphenyl)ethan-1-one, 1-(3-Bromo-2-hydroxyphenyl)ethanone, 2-Acetyl-6-bromophenol
OR54360-1G OR54360-5G OR54360-25G
APOSOR54360-1GEA 39.3 EUR
APOSOR54360-1G APOSOR54360-5G APOSOR54360-25G
3'-Bromo-2'-hydroxyacetophenone 98%
3'-Bromo-2'-hydroxyacetophenone
Formula: C₈H₇BrO₂
MW: 215.044 g/mol
Boiling Pt: 140…145 °C (6 mm Hg)
Melting Pt: 40…44 °C
Storage Temperature: Ambient
MDL Number: MFCD08236725
CAS Number: 1836-05-1