Pent-4-enoic anhydride 98%
Supplier: Apollo Scientific
Warning
OR54404-25ML OR54404-5ML
APOSOR54404-25MLEA 546 EUR
APOSOR54404-25ML APOSOR54404-5ML
Pent-4-enoic anhydride 98%
Pent-4-enoic anhydride
Formula: C₁₀H₁₄O₃
Boiling Pt: 78…81 °C (0.375 mm Hg)
Density: 0.997 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01320671
CAS Number: 63521-92-6