N,N'-bis(2-Phenylphenyl)ethanediamide 95%

Supplier: Apollo Scientific
Warning

Synonyms: Ethanediamide, N1,N2-bis([1,1'-biphenyl]-2-yl)-

OR54683-1G OR54683-5G
APOSOR54683-1GEA 21.9 EUR
APOSOR54683-1G APOSOR54683-5G
N,N'-bis(2-Phenylphenyl)ethanediamide 95%
N,N'-bis(2-Phenylphenyl)ethanediamide
Formula: C₂₆H₂₀N₂O₂
MW: 392.449 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31793560
CAS Number: 21022-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR