2,5-Dimethylphenacyl bromide
Supplier: Apollo Scientific
Danger
OR54697-1G OR54697-5G
APOSOR54697-1GEA 125 EUR
APOSOR54697-1G APOSOR54697-5G
2,5-Dimethylphenacyl bromide
2,5-Dimethylphenacyl bromide
Formula: C₁₀H₁₁BrO
MW: 227.098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07364273
CAS Number: 75840-13-0
UN: 3261
ADR: 8 III