4-(2-Hydroxyethyl)-2-methoxyphenol 99%
Supplier: Apollo Scientific
Warning
OR55020-1G OR55020-250MG
APOSOR55020-1GEA 143 EUR
APOSOR55020-1G APOSOR55020-250MG
4-(2-Hydroxyethyl)-2-methoxyphenol 99%
4-(2-Hydroxyethyl)-2-methoxyphenol
Formula: C₉H₁₂O₃
Melting Pt: 40…42 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD0002903
CAS Number: 2380-78-1