3-Methoxyoxan-4-one
Supplier: Apollo Scientific
Synonyms: 3-Methoxy-tetrahydropyran-4-one, 3-Methoxydihydro-2H-pyran-4(3H)-one
OR55049-1G OR55049-5G
APOSOR55049-1GEA 111 EUR
APOSOR55049-1G APOSOR55049-5G
3-Methoxyoxan-4-one
3-Methoxyoxan-4-one
Formula: C₆H₁₀O₃
MW: 130.142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09909903
CAS Number: 624734-17-4