2,4-Dichloro-5-(methylsulphanyl)pyrimidine
Supplier: Apollo Scientific
Danger
Synonyms: 2,4-Dichloro-5-(methylthio)pyrimidine
OR55092-1G OR55092-5G
APOSOR55092-1GEA 120 EUR
APOSOR55092-1G APOSOR55092-5G
2,4-Dichloro-5-(methylsulphanyl)pyrimidine
2,4-Dichloro-5-(methylsulphanyl)pyrimidine
Formula: C₅H₄Cl₂N₂S
MW: 195.07 g/mol
Melting Pt: 105…106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22199809
CAS Number: 7401-98-1