2-Methoxy-4,6-dimethylbenzeneboronic acid
Supplier: Apollo Scientific
Warning
Synonyms: (2-Methoxy-4,6-dimethylphenyl)boronic acid
OR55266-1G
APOSOR55266-1GEA 127 EUR
APOSOR55266-1G
2-Methoxy-4,6-dimethylbenzeneboronic acid
2-Methoxy-4,6-dimethylbenzeneboronic acid
Formula: C₉H₁₃BO₃
MW: 180.009 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04037222
CAS Number: 355836-08-7