tert-Butyl 4-formyl-1H-pyrazole-1-carboxylate
Supplier: Apollo Scientific
Synonyms: tert-Butyl 4-formylpyrazole-1-carboxylate
OR55293-1G OR55293-5G
APOSOR55293-1GEA 98.6 EUR
APOSOR55293-1G APOSOR55293-5G
tert-Butyl 4-formyl-1H-pyrazole-1-carboxylate
tert-Butyl 4-formyl-1H-pyrazole-1-carboxylate
Formula: C₉H₁₂N₂O₃
MW: 196.203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19707636
CAS Number: 821767-61-7