tert-Butyl 4-bromo-5,6-dihydropyridine-1(2H)-carboxylate 95%
Supplier: Apollo Scientific
Synonyms: tert-Butyl 4-bromo-1,2,3,6-tetrahydropyridine-1-carboxylate, tert-Butyl 4-bromo-3,6-dihydro-2H-pyridine-1-carboxylate
OR55294-1G OR55294-5G
APOSOR55294-1GEA 79.9 EUR
APOSOR55294-1G APOSOR55294-5G
tert-Butyl 4-bromo-5,6-dihydropyridine-1(2H)-carboxylate 95%
tert-Butyl 4-bromo-5,6-dihydropyridine-1(2H)-carboxylate
Formula: C₁₀H₁₆BrNO₂
MW: 262.144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13181893
CAS Number: 159503-91-0