5-Bromo-4-methoxy-2-methylaniline 95+%
Supplier: Apollo Scientific
OR55314-250MG
OR55314-1G
APOSOR55314-250MGEA
62.5
EUR
APOSOR55314-250MG
APOSOR55314-1G
5-Bromo-4-methoxy-2-methylaniline 95+%
5-Bromo-4-methoxy-2-methylaniline
Formula:
C₈H₁₀BrNO MW: 216.075 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17392823 CAS Number: 861084-04-0 |