3-(4-Bromophenoxy)-N,N-dimethylpropylamine 97%
Supplier: Apollo Scientific
OR55320-1G OR55320-5G
APOSOR55320-1GEA 125 EUR
APOSOR55320-1G APOSOR55320-5G
3-(4-Bromophenoxy)-N,N-dimethylpropylamine 97%
3-(4-Bromophenoxy)-N,N-dimethylpropylamine
Formula: C₁₁H₁₆BrNO
MW: 258.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03718813
CAS Number: 76579-64-1