2-Bromo-6-chloro-1,8-naphthyridine 95%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-6-chloro-1,8-naphthyridine
OR55345-100MG OR55345-250MG
APOSOR55345-100MGEA 836 EUR
APOSOR55345-100MG APOSOR55345-250MG
2-Bromo-6-chloro-1,8-naphthyridine 95%
2-Bromo-6-chloro-1,8-naphthyridine
Formula: C₈H₄BrClN₂
MW: 243.488 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18253968
CAS Number: 1176470-56-6