cis-1-Bromo-3-methoxycyclobutane 95%
Supplier: Apollo Scientific
Warning
OR55390-100MG OR55390-250MG
APOSOR55390-100MGEA 0 EUR
APOSOR55390-100MG APOSOR55390-250MG
cis-1-Bromo-3-methoxycyclobutane 95%
cis-1-Bromo-3-methoxycyclobutane
Formula: C₅H₉BrO
MW: 165.028 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30487877
CAS Number: 2044705-32-8