8-Nitro-1,2,3,9b-tetrahydrobenzo[c]thieno[2,1-e]isothiazole 4-oxide 95%
Supplier: Apollo Scientific
Warning
OR55437-1G OR55437-250MG
APOSOR55437-1GEA 1010 EUR
APOSOR55437-1G APOSOR55437-250MG
8-Nitro-1,2,3,9b-tetrahydrobenzo[c]thieno[2,1-e]isothiazole 4-oxide 95%
8-Nitro-1,2,3,9b-tetrahydrobenzo[c]thieno[2,1-e]isothiazole 4-oxide
Formula: C₁₀H₁₀N₂O₃S
MW: 238.263 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31629919
CAS Number: 2169311-00-4