Methyl 3-bromo-5-chlorobenzoate 98%
Supplier: Apollo Scientific
Warning
OR55474-1G OR55474-5G
APOSOR55474-1GEA 156 EUR
APOSOR55474-1G APOSOR55474-5G
Methyl 3-bromo-5-chlorobenzoate 98%
Methyl 3-bromo-5-chlorobenzoate
Formula: C₈H₆BrClO₂
MW: 249.489 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07780734
CAS Number: 933585-58-1