(1R,3R,5S)-9-Benzyl-9-azabicyclo[3.3.1]nonan-3-ol
Supplier: Apollo Scientific
Warning
Synonyms: Endo-9-benzyl-9-azabicyclo[3.3.1]nonan-3-ol, (3-endo)-9-Benzyl-9-azabicyclo[3.3.1]nonan-3-ol
OR55481-1G OR55481-250MG
APOSOR55481-1GEA 1020 EUR
APOSOR55481-1G APOSOR55481-250MG
(1R,3R,5S)-9-Benzyl-9-azabicyclo[3.3.1]nonan-3-ol
(1R,3R,5S)-9-Benzyl-9-azabicyclo[3.3.1]nonan-3-ol
Formula: C₁₅H₂₁NO
MW: 231.333 g/mol
Melting Pt: 69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00209918
CAS Number: 70243-51-5