(S)-3-(Bromomethyl)-1-methylpiperidine hydrobromide
Supplier: Apollo Scientific
Danger
OR55484-1G OR55484-250MG OR55484-5G
APOSOR55484-1GEA 364 EUR
APOSOR55484-1G APOSOR55484-250MG APOSOR55484-5G
(S)-3-(Bromomethyl)-1-methylpiperidine hydrobromide
(S)-3-(Bromomethyl)-1-methylpiperidine hydrobromide
Formula: C₇H₁₄BrN·BrH
MW: 273.009 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31629912
CAS Number: 2244064-27-3
UN: 1759
ADR: 8,II