1-Hydroxy-4-methylphthalazine 98%
Supplier: Apollo Scientific
Warning
OR55544-1G OR55544-5G OR55544-10G
APOSOR55544-1GEA 89.9 EUR
APOSOR55544-1G APOSOR55544-5G APOSOR55544-10G
1-Hydroxy-4-methylphthalazine 98%
1-Hydroxy-4-methylphthalazine
Formula: C₉H₈N₂O
MW: 160.173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234527
CAS Number: 5004-48-8