2-(tert-Butoxy)-2-oxoethylzinc bromide 0.5M in diethyl ether
Supplier: Apollo Scientific
Danger
Synonyms: Bromo-[2-[(2-methylpropan-2-yl)oxy]-2-oxoethyl]zinc, (2-tert-Butoxy-2-oxoethyl)zinc(II) bromide
OR55579-100ML OR55579-50ML
APOSOR55579-100MLEA 1470 EUR
APOSOR55579-100ML APOSOR55579-50ML
2-(tert-Butoxy)-2-oxoethylzinc bromide 0.5M in diethyl ether
2-(tert-Butoxy)-2-oxoethylzinc bromide
Formula: C₆H₁₁BrO₂Zn
MW: 260.463 g/mol
Flash Pt: –40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00671971
CAS Number: 51656-70-3
UN: 1993
ADR: 3 I