1-Methyl-7-nitroisatoic anhydride 95%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Methyl-7-nitro-3,1-benzoxazine-2,4-dione
OR55588-1G OR55588-250MG OR55588-5G
APOSOR55588-1GEA 372 EUR
APOSOR55588-1G APOSOR55588-250MG APOSOR55588-5G
1-Methyl-7-nitroisatoic anhydride 95%
1-Methyl-7-nitroisatoic anhydride
Formula: C₉H₆N₂O₅
MW: 222.154 g/mol
Melting Pt: 203…206 °C
Storage Temperature: Ambient
MDL Number: MFCD18432729
CAS Number: 73043-80-8