4-(But-1-yl)phenacyl bromide
Supplier: Apollo Scientific
Danger
OR55607-1G OR55607-25G OR55607-5G
APOSOR55607-1GEA 56.7 EUR
APOSOR55607-1G APOSOR55607-25G APOSOR55607-5G
4-(But-1-yl)phenacyl bromide
4-(But-1-yl)phenacyl bromide
Formula: C₁₂H₁₅BrO
MW: 255.151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11182347
CAS Number: 64356-03-2
UN: 3261
ADR: 8,III