2,6-Dibromobenzenesulphonyl chloride
Supplier: Apollo Scientific
Danger
OR55628-10G OR55628-1G OR55628-5G
APOSOR55628-10GEA 509 EUR
APOSOR55628-10G APOSOR55628-1G APOSOR55628-5G
2,6-Dibromobenzenesulphonyl chloride
2,6-Dibromobenzenesulphonyl chloride
Formula: C₆H₃Br₂ClO₂S
MW: 334.413 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226510
CAS Number: 184170-43-2
UN: 3261
ADR: 8,III