1,5-Dimethyl-4-bromopyrazole-3-carbonitrile
Supplier: Apollo Scientific
Danger
OR55637-1G OR55637-250MG
APOSOR55637-1GEA 574 EUR
APOSOR55637-1G APOSOR55637-250MG
1,5-Dimethyl-4-bromopyrazole-3-carbonitrile
1,5-Dimethyl-4-bromopyrazole-3-carbonitrile
Formula: C₆H₆BrN₃
MW: 200.036 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30478437
CAS Number: 863751-81-9
UN: 3439
ADR: 6.1 III