(4R)-4-(Aminomethyl)pyrrolidin-2-one 97%
Supplier: Apollo Scientific
Warning
OR55658-1G OR55658-250MG
APOSOR55658-1GEA 1740 EUR
APOSOR55658-1G APOSOR55658-250MG
(4R)-4-(Aminomethyl)pyrrolidin-2-one 97%
(4R)-4-(Aminomethyl)pyrrolidin-2-one
Formula: C₅H₁₀N₂O
MW: 114.146 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23106121
CAS Number: 1407997-81-2