4,6-Dibromo-2-chloroanisole
Supplier: Apollo Scientific
Warning
OR55675-1G OR55675-500MG
APOSOR55675-1GEA 357 EUR
APOSOR55675-1G APOSOR55675-500MG
4,6-Dibromo-2-chloroanisole
4,6-Dibromo-2-chloroanisole
Formula: C₇H₅Br₂ClO
MW: 300.375 g/mol
Boiling Pt: 269…272 °C
Melting Pt: 75…76 °C
Density: 2.213 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD22543700
CAS Number: 174913-39-4